Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pdf-1
Entrez Gene ID
175963
Gene Name
protein-coding
Gene Aliases
CELE_T07E3.6
Chromosome Location
Chr.III: 6907410 - 6912481 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001382903.1 1 - 2 369 86

View Full Product Details