Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cyk-1
Entrez Gene ID
175983
Gene Name
protein-coding
Gene Aliases
CELE_F11H8.4
Chromosome Location
Chr.III: 7023642 - 7029205 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_171180.5 NP_741210.3 10 - 11 2692 102
NM_171880.6 NP_741211.3 10 - 11 2694 102

View Full Product Details