Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
unc-16
Entrez Gene ID
176349
Gene Name
protein-coding
Gene Aliases
CELE_ZK1098.10
Chromosome Location
Chr.III: 9546258 - 9557450 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027810.5 NP_001022981.1 16 - 17 2698 94
NM_001129275.4 NP_001122747.1 6 - 7 801 94
NM_001129276.3 NP_001122748.1 17 - 18 2782 94
NM_001268079.4 NP_001255008.1 17 - 18 2797 94
NM_001268080.3 NP_001255009.1 12 - 13 1887 94
NM_001306679.3 NP_001293608.1 10 - 11 1683 94
NM_001306687.3 NP_001293616.1 11 - 12 1701 94
NM_001306688.3 NP_001293617.1 7 - 8 885 94
NM_001382965.1 19 - 20 3103 94
NM_171221.7 NP_741263.3 18 - 19 2789 94
GenBank mRNA AF424978.1 - 16 - 17 2634 94

View Full Product Details