Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
unc-16
Entrez Gene ID
176349
Gene Name
protein-coding
Gene Aliases
CELE_ZK1098.10
Chromosome Location
Chr.III: 9546258 - 9557450 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027810.5 NP_001022981.1 4 - 5 722 125
NM_001129276.3 NP_001122748.1 4 - 5 722 125
NM_001268079.4 NP_001255008.1 4 - 5 722 125
NM_001306680.3 NP_001293609.1 4 - 5 658 125
NM_001306682.3 NP_001293611.1 3 - 4 550 125
NM_001306684.3 NP_001293613.1 4 - 5 658 125
NM_001306686.3 NP_001293615.1 4 - 5 658 125
NM_001382965.1 4 - 5 722 125
NM_171221.7 NP_741263.3 4 - 5 759 125
GenBank mRNA AF424978.1 - 4 - 5 658 125

View Full Product Details