Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tra-1
Entrez Gene ID
176548
Gene Name
protein-coding
Gene Aliases
CELE_Y47D3A.6
Chromosome Location
Chr.III: 11170889 - 11195207 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027709.3 NP_001022880.2 7 - 8 810 71
NM_001027710.6 NP_001022881.2 7 - 8 804 71
GenBank mRNA M93256.1 - 7 - 8 825 71
M94130.1 - 7 - 8 825 71

View Full Product Details