Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
unc-64
Entrez Gene ID
176743
Gene Name
protein-coding
Gene Aliases
CELE_F56A8.7
Chromosome Location
Chr.III: 13277259 - 13283887 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027443.5 NP_001022614.1 7 - 8 894 73
GenBank mRNA AB008843.1 - 7 - 8 904 73
AB008844.1 - 7 - 8 904 73

View Full Product Details