Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
prdx-6
Entrez Gene ID
176837
Gene Name
Thioredoxin domain-containing protein
Gene Aliases
CELE_Y38C1AA.11
Chromosome Location
Chr.IV: 218551 - 220855 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_171241.4 NP_741287.1 1 - 2 106 105

View Full Product Details