Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
osm-3
Entrez Gene ID
177141
Gene Name
protein-coding
Gene Aliases
CELE_M02B7.3
Chromosome Location
Chr.IV: 3796300 - 3800461 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001380163.1 NP_001367796.1 9 - 10 1957 110
NM_001380164.2 NP_001367795.1 9 - 10 1971 110

View Full Product Details