Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
skn-1
Entrez Gene ID
177343
Gene Name
protein-coding
Gene Aliases
CELE_T19E7.2
Chromosome Location
Chr.IV: 5651039 - 5660384 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001380237.1 NP_001367379.1 4 - 5 1162 90
NM_001380238.1 NP_001368358.1 1 - 2 232 90
NM_171345.6 NP_741404.1 6 - 7 1432 90
NM_171347.7 NP_741406.1 2 - 3 627 90

View Full Product Details