The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001306814.4 | NP_001293743.1 | 3 - 4 | 402 | 94 |
NM_001306815.3 | NP_001293744.1 | 3 - 4 | 402 | 94 | |
NM_068703.7 | NP_501104.2 | 2 - 3 | 162 | 94 | |
NM_171374.6 | NP_741437.1 | 2 - 3 | 162 | 94 |