Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nhr-8
Entrez Gene ID
177551
Gene Name
protein-coding
Gene Aliases
CELE_F33D4.1
Chromosome Location
Chr.IV: 7710975 - 7714210 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_171382.4 NP_741445.1 4 - 5 737 78
GenBank mRNA AF083226.1 - 4 - 5 725 78

View Full Product Details