Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
unc-94
Entrez Gene ID
172275
Gene Name
protein-coding
Gene Aliases
CELE_C06A5.7
Chromosome Location
Chr.I: 5992659 - 6007600 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059334.5 NP_491735.1 1 - 2 198 72

View Full Product Details