Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rnf-1
Entrez Gene ID
172277
Gene Name
RING-type domain-containing protein
Gene Aliases
CELE_C06A5.9
Chromosome Location
Chr.I: 6009782 - 6011586 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059337.5 NP_491738.2 7 - 8 910 63

View Full Product Details