Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
vps-34
Entrez Gene ID
172280
Gene Name
protein-coding
Gene Aliases
CELE_B0025.1
Chromosome Location
Chr.I: 6028482 - 6033458 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001380450.1 NP_001367304.1 2 - 3 749 108
NM_059340.5 NP_491741.1 2 - 3 761 108

View Full Product Details