Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
haf-9
Entrez Gene ID
172288
Gene Name
protein-coding
Gene Aliases
CELE_ZK484.2
Chromosome Location
Chr.I: 6081782 - 6086006 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059353.7 NP_491754.1 13 - 14 2003 122

View Full Product Details