Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
oac-9
Entrez Gene ID
172323
Gene Name
NRF domain-containing protein
Gene Aliases
CELE_C48E7.8
Chromosome Location
Chr.I: 6265678 - 6269073 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059408.5 NP_491809.2 7 - 8 1525 139

View Full Product Details