Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ima-2
Entrez Gene ID
172329
Gene Name
Importin subunit alpha-2
Gene Aliases
CELE_F26B1.3
Chromosome Location
Chr.I: 6315098 - 6317267 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059423.7 NP_491824.1 3 - 4 486 61
GenBank mRNA AF040996.1 - 2 - 3 444 61
AF326936.1 - 2 - 3 448 61

View Full Product Details