Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
bli-4
Entrez Gene ID
172333
Gene Name
protein-coding
Gene Aliases
CELE_K04F10.4
Chromosome Location
Chr.I: 6341311 - 6356680 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026372.7 NP_001021543.1 17 - 18 2882 97

View Full Product Details