Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ncbp-1
Entrez Gene ID
172345
Gene Name
Nuclear cap-binding protein subunit 1
Gene Aliases
CELE_F37E3.1
Chromosome Location
Chr.I: 6421978 - 6425794 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059449.6 NP_491850.2 4 - 5 1034 85

View Full Product Details