Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
npp-12
Entrez Gene ID
172348
Gene Name
Nuclear Pore complex Protein
Gene Aliases
CELE_T23H2.1
Chromosome Location
Chr.I: 6447304 - 6453933 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059454.4 NP_491855.1 2 - 3 337 74

View Full Product Details