Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mdt-18
Entrez Gene ID
183844
Gene Name
protein-coding
Gene Aliases
CELE_C55B7.9
Chromosome Location
Chr.I: 6492722 - 6494781 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383258.1 3 - 4 191 92
NM_059468.6 NP_491869.1 3 - 4 226 92

View Full Product Details