Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
C55B7.3
Entrez Gene ID
172359
Gene Name
Tyrosine-protein phosphatase
Gene Aliases
CELE_C55B7.3
Chromosome Location
Chr.I: 6497450 - 6501413 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001380558.1 NP_001367695.1 3 - 4 728 64

Gene Symbol
acdh-1
Entrez Gene ID
172358
Gene Name
protein-coding
Gene Aliases
CELE_C55B7.4
Chromosome Location
Chr.I: 6497436 - 6501413 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001380557.1 NP_001367696.1 1 - 2 446 64
NM_001383261.1 3 - 4 742 64

View Full Product Details