Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
glh-2
Entrez Gene ID
172361
Gene Name
ATP-dependent RNA helicase glh-2
Gene Aliases
CELE_C55B7.1
Chromosome Location
Chr.I: 6513928 - 6517507 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059475.6 NP_491876.1 4 - 5 1197 95

View Full Product Details