Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
try-6
Entrez Gene ID
185959
Gene Name
Peptidase S1 domain-containing protein
Gene Aliases
CELE_F48A9.3
Chromosome Location
Chr.I: 6580186 - 6587902 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001374905.2 NP_001361988.1 - 3168 79

View Full Product Details