Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ppfr-2
Entrez Gene ID
172381
Gene Name
Protein Phosphatase Four Regulatory subunit
Gene Aliases
CELE_D2092.2
Chromosome Location
Chr.I: 6625042 - 6626788 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059506.6 NP_491907.1 2 - 3 305 87

View Full Product Details