Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mec-6
Entrez Gene ID
172389
Gene Name
Mechanosensory abnormality protein 6
Gene Aliases
CELE_W02D3.3
Chromosome Location
Chr.I: 6724889 - 6728330 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383275.1 4 - 5 466 127
GenBank mRNA AF295927.1 - 3 - 4 364 127

View Full Product Details