Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- dhod-1
- Entrez Gene ID
-
172392
- Gene Name
- Dihydroorotate dehydrogenase (quinone), mitochondrial
- Gene Aliases
-
CELE_W02D3.2
- Chromosome Location
-
Chr.I: 6736438 - 6741176 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_059529.7 |
NP_491930.1 |
5 - 6 |
1328 |
88 |
- Gene Symbol
- cytb-5.2
- Entrez Gene ID
-
172393
- Gene Name
- Cytochrome b5 heme-binding domain-containing protein
- Gene Aliases
-
CELE_W02D3.1
- Chromosome Location
-
Chr.I: 6736677 - 6741889 on Build WBcel235
View Full Product Details