Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fnci-1
Entrez Gene ID
189113
Gene Name
FANCI_HD2 domain-containing protein
Gene Aliases
CELE_W02D3.10
Chromosome Location
Chr.I: 6745988 - 6753031 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026481.5 NP_001021652.1 3 - 4 1092 84

View Full Product Details