Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
acdh-5
Entrez Gene ID
172400
Gene Name
Acyl CoA DeHydrogenase
Gene Aliases
CELE_C37A2.3
Chromosome Location
Chr.I: 6784854 - 6787139 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059541.4 NP_491942.1 4 - 5 878 87

View Full Product Details