Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dpf-3
Entrez Gene ID
172409
Gene Name
protein-coding
Gene Aliases
CELE_K02F2.1
Chromosome Location
Chr.I: 6838354 - 6843343 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059555.2 NP_491956.1 17 - 18 2403 113
NM_059556.5 NP_491957.1 17 - 18 2391 113

View Full Product Details