Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
glh-1
Entrez Gene ID
172414
Gene Name
ATP-dependent RNA helicase glh-1
Gene Aliases
CELE_T21G5.3
Chromosome Location
Chr.I: 6854478 - 6857536 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059562.6 NP_491963.1 1 - 2 371 59

View Full Product Details