Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
asd-2
Entrez Gene ID
188703
Gene Name
protein-coding
Gene Aliases
CELE_T21G5.5
Chromosome Location
Chr.I: 6877059 - 6892143 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026454.5 NP_001021625.1 1 - 2 178 64
NM_001026455.6 NP_001021626.1 3 - 4 338 64
NM_001263411.2 NP_001250340.1 3 - 4 338 64
GenBank mRNA EF630625.1 - 1 - 2 34 64

View Full Product Details