Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
srb-15
Entrez Gene ID
183571
Gene Name
Serpentine receptor class beta-15
Gene Aliases
CELE_C48B6.5
Chromosome Location
Chr.I: 6897735 - 6900619 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059568.3 NP_491969.2 2 - 3 230 149

View Full Product Details