Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ztf-4
Entrez Gene ID
172422
Gene Name
RRM domain-containing protein
Gene Aliases
CELE_T10B11.3
Chromosome Location
Chr.I: 6926257 - 6929665 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059575.5 NP_491976.2 4 - 5 404 93

View Full Product Details