Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sem-4
Entrez Gene ID
172435
Gene Name
Uncharacterized protein
Gene Aliases
CELE_F15C11.1
Chromosome Location
Chr.I: 7038900 - 7048832 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059596.5 NP_491997.1 4 - 5 450 105
GenBank mRNA U60112.1 - 4 - 5 450 105

View Full Product Details