Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
eat-5
Entrez Gene ID
172480
Gene Name
Innexin eat-5
Gene Aliases
CELE_F13G3.8
Chromosome Location
Chr.I: 7308643 - 7311939 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059667.7 NP_492068.1 6 - 7 1148 82
GenBank mRNA U59210.1 - 6 - 7 1125 82

View Full Product Details