Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
glb-18
Entrez Gene ID
172485
Gene Name
protein-coding
Gene Aliases
CELE_F52A8.4
Chromosome Location
Chr.I: 7339232 - 7345986 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001136303.4 NP_001129775.1 5 - 6 551 73
NM_059673.7 NP_492074.1 5 - 6 610 73

View Full Product Details