Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
inx-14
Entrez Gene ID
172488
Gene Name
protein-coding
Gene Aliases
CELE_F07A5.1
Chromosome Location
Chr.I: 7350309 - 7352235 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059677.7 NP_492078.1 5 - 6 1037 143
NM_059678.6 NP_492079.1 5 - 6 1019 143

View Full Product Details