Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
unc-15
Entrez Gene ID
172491
Gene Name
protein-coding
Gene Aliases
CELE_F07A5.7
Chromosome Location
Chr.I: 7357240 - 7383197 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059684.6 NP_492085.1 6 - 7 762 77

View Full Product Details