Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sol-1
Entrez Gene ID
172493
Gene Name
Suppressor of lurcher protein 1
Gene Aliases
CELE_C15A11.3
Chromosome Location
Chr.I: 7389049 - 7395918 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059687.7 NP_492088.3 10 - 11 1338 148
GenBank mRNA AY387697.1 - 10 - 11 1303 148

View Full Product Details