Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
asic-2
Entrez Gene ID
189058
Gene Name
Degenerin-like protein asic-2
Gene Aliases
CELE_T28F4.2
Chromosome Location
Chr.I: 7480108 - 7483505 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059698.3 NP_492099.1 10 - 11 1364 108

View Full Product Details