Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nas-36
Entrez Gene ID
172506
Gene Name
Zinc metalloproteinase nas-36
Gene Aliases
CELE_C26C6.3
Chromosome Location
Chr.I: 7527224 - 7531715 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059708.4 NP_492109.2 10 - 11 1176 115

View Full Product Details