Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
wdr-23
Entrez Gene ID
172518
Gene Name
protein-coding
Gene Aliases
CELE_D2030.9
Chromosome Location
Chr.I: 7596505 - 7602221 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001263616.4 NP_001250545.1 9 - 10 1510 64
NM_001306353.3 NP_001293282.1 7 - 8 1294 64
NM_001306354.3 NP_001293283.1 7 - 8 1270 64
NM_059723.7 NP_492124.1 9 - 10 1532 64
NM_059724.4 NP_492125.1 8 - 9 1550 64

Gene Symbol
D2030.8
Entrez Gene ID
172517
Gene Name
Uncharacterized protein
Gene Aliases
CELE_D2030.8
Chromosome Location
Chr.I: 7591288 - 7602206 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059722.6 NP_492123.2 18 - 19 4859 64

View Full Product Details