Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
del-3
Entrez Gene ID
184955
Gene Name
DEgenerin Like
Gene Aliases
CELE_F26A3.6
Chromosome Location
Chr.I: 7670152 - 7672714 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059734.3 NP_492135.1 6 - 7 1306 138

View Full Product Details