Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rlbp-1
Entrez Gene ID
172530
Gene Name
protein-coding
Gene Aliases
CELE_T23G11.5
Chromosome Location
Chr.I: 7683341 - 7694210 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001263636.3 NP_001250565.1 5 - 6 626 138
NM_001267076.2 NP_001254005.1 8 - 9 1259 138

View Full Product Details