Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gld-1
Entrez Gene ID
172532
Gene Name
Female germline-specific tumor suppressor gld-1
Gene Aliases
CELE_T23G11.3
Chromosome Location
Chr.I: 7695661 - 7698227 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059742.7 NP_492143.1 1 - 2 218 60

View Full Product Details