Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
unc-55
Entrez Gene ID
192082
Gene Name
protein-coding
Gene Aliases
CELE_F55D12.4
Chromosome Location
Chr.I: 7898434 - 7903334 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001129010.3 NP_001122482.2 6 - 7 927 152
NM_001129011.4 NP_001122483.2 6 - 7 888 152
GenBank mRNA EF601077.1 - 6 - 7 888 152
EF601082.1 - 6 - 7 927 152

View Full Product Details