Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rbg-2
Entrez Gene ID
172574
Gene Name
protein-coding
Gene Aliases
CELE_T22C1.10
Chromosome Location
Chr.I: 7962052 - 7972450 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026460.3 NP_001021631.1 8 - 9 2801 116
NM_001026461.4 NP_001021632.1 8 - 9 2799 116

View Full Product Details