Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
che-3
Entrez Gene ID
172593
Gene Name
Cytoplasmic dynein 2 heavy chain 1
Gene Aliases
CELE_F18C12.1
Chromosome Location
Chr.I: 8062007 - 8081671 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059820.4 NP_492221.2 1 - 2 161 115

View Full Product Details