Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lin-10
Entrez Gene ID
172597
Gene Name
protein-coding
Gene Aliases
CELE_C09H6.2
Chromosome Location
Chr.I: 8109746 - 8119318 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025825.5 NP_001020996.1 12 - 13 2254 102
NM_001136277.4 NP_001129749.1 12 - 13 2182 102
NM_059825.7 NP_492226.2 13 - 14 2340 102
GenBank mRNA AJ133374.1 - 12 - 13 2251 102

View Full Product Details